Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Gas Detection Systems
Single-Gas Monitor
Single-Gas Monitor
BWTEBWC3R-M30200
:
BW TECHNOLOGIES
New Product
Single-Gas Monitor
BWTEBWC3R-M30200
:
BW TECHNOLOGIES
New Product
Model:
BW Clip RT
Parameter(s):
CO
Description:
Single gas detector for Carbon monoxide (CO) 30 - 200 ppm
Pk:
1