Order Entry
Austria
ContactUsLinkComponent
Brady® ToughStripe® Max Floor Marking Tapes
Brady® ToughStripe® Max Floor Marking Tapes
Artikelnr. BRDY177017
Supplier:  Brady
New Product
undefined
Brady® ToughStripe® Max Floor Marking Tapes
Artikelnr. BRDY177017
Supplier:  Brady
New Product

Specifications

  • Width:
    50,8 mm
  • Colour:
    Yellow/black
  • Description:
    ToughStripe® Max floor marking tape, hazard stripes finish
  • Length:
    30,48 m
  • Thickness:
    0,61 mm
  • Pk:
    1

Specifications

About this item

Built stronger to last longer on punishing industrial floors. ToughStripe® Max floor marking tape is purpose-built to hold its ground in forklift and heavy traffic areas.

  • Permanent rubber based adhesive
  • Designed for indoor use
  • Gloss finish

50% thinner than previous version of ToughStripe® Max and twice as tough as similarly thick vinyl tapes. Thinner profile and true edge-to-edge adhesive minimise pick points. Plasticizer-free polyester (PETG) material is more rigid than vinyl. Every layer is hand selected, pressure tested, and purpose built to protect colours and outlast the competition. Damage-isolating design keeps tape intact longer by preventing rips, gouges, and tears from running. Tape provides excellent colour contrast for clear visibility under any lighting. Compact construction results in a smaller, lightweight roll. Can be easily installed by one person with minimal tools and is compatible with the ToughStripe® floor tape applicator.