ContactUsLinkComponent
PCA - Plate Count Agar
PCA - Plate Count Agar
Catalog # DOEH876631782
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
PCA - Plate Count Agar
Catalog # DOEH876631782
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    9
  • Description:
    Plate count agar (PCA)
  • Size:
    9×250 ml/bottle
Specifications
About this item
Agar medium for the quantitative detection of the total germ count in the beverage and food industry.
  • Ready-to-use, liquefaction possible directly in the bottle
  • Universal applicable (membrane filtration, pouring and spread plate method)
  • Reference medium, pH neutral
The number of detectable microorganisms is an important indicator for quality assurance. The medium is also known as Bacteria Count Agar or Caseine-peptone Dextrose Yeast Agar and complies with the requirements of EN ISO 4833 and ISO 17410.