Order Entry
Austria
ContactUsLinkComponent
Specpure® Multi Ion Anion IC Standard Solution in H₂O
Specpure® Multi Ion Anion IC Standard Solution in H₂O
Catalog # 41694.AE
CAS Number:  
Specpure® Multi Ion Anion IC Standard Solution in H₂O
Catalog # 41694.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Multi Ion anion IC standard solution in H₂O
  • MDL No.:
    MFCD00144813
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00144813