Order Entry
Austria
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Etofenprox-(Ethoxy-D₅)
Supelco® PESTANAL® Organic Reference Standard, Etofenprox-(Ethoxy-D₅)
  34513-10MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Etofenprox-(Ethoxy-D₅)
  34513-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1705649-55-3
  • Description:
    Etofenprox-(ethoxy-d₅)
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H362 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD11616821
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-(4-Ethoxy-d₅-phenyl)-2-methylpropyl 3-phenoxybenzyl ether; Etofenprox-d₅
  • Formula:
    C₂₅D₅H₂₃O₃
  • Molecular weight:
    381,52
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS09
 : P201 - P260 - P263 - P273 - P308 + P313 - P391