Supelco® PESTANAL® Organic Reference Standard, Atrazine
Catalog # 45330-250MG-R
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1912-24-9
- Description:Atrazine
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00041810
- M-Clarity quality level:MQ100
- Synonym(s):2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
- Formula:C₈H₁₄ClN₅
- Molecular weight:215,68
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09