ContactUsLinkComponent
SO Green™ 520WS Singlet Oxygen Sensor *Water-Soluble for Extracellular Applications*
SO Green™ 520WS Singlet Oxygen Sensor *Water-Soluble for Extracellular Applications*
Catalog # AATB16061
Supplier:  AAT Bioquest
New Product
CAS Number:  
undefined
SO Green™ 520WS Singlet Oxygen Sensor *Water-Soluble for Extracellular Applications*
Catalog # AATB16061
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Description:
    SO Green™ 520WS Singlet Oxygen sensor *Water-soluble for extracellular applications*, 10×100 μg
Specifications
About this item
SO Green™ 520WS Singlet Oxygen Sensor has been developed as a water-soluble fluorescent probe for detecting extracellular singlet oxygen since it is cell-impermeant.
  • Stable for 24 months
Quality tested in-house.
Caution: For research use only.