ContactUsLinkComponent
HIS Lite™ Cy3 Bis NTA Chelator
HIS Lite™ Cy3 Bis NTA Chelator
Catalog # AATB12650
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
HIS Lite™ Cy3 Bis NTA Chelator
Catalog # AATB12650
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Description:
    HIS Lite™ Cy3 Bis NTA chelator
Specifications
About this item
Cy3 Bis NTA-Ni Chelator can be applied either to an SDS-PAGE gel or Western blot membrane for fluorescence imaging or used in living cells in combination with the addition of a heavy metal ion such as Ni²⁺.
  • Stable for 24 months
Quality tested in-house.
Caution: For research use only.