Order Entry
Austria
ContactUsLinkComponent
Magicard™ Colour Film + Transfer Tape Set for Prima 8
Magicard™ Colour Film + Transfer Tape Set for Prima 8
Catalog # BRDY321977
Supplier:  Brady
New Product
undefined
Magicard™ Colour Film + Transfer Tape Set for Prima 8
Catalog # BRDY321977
Supplier:  Brady
New Product

Specifications

  • Colour:
    Assorted
  • Description:
    YMCK dye film + re-transfer film set, 4-colour ribbon (Cyan, Magenta, Yellow, Black), 1000 single-sided cards
  • Length:
    9,75 m
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Prima 8

Specifications

About this item

Set of dye film and transfer film all in one.

  • Print full colour on one side and black on the other
  • Print 1000 cards front and back in one go

Dye film and re-transfer film that fits the Prima 8 printers.

Delivery: Includes 1 YMCK dye film and 1 re-transfer film.