ReadiLink™ Rapid XFD555 Antibody Labelling Kit
Catalog # AATB5733
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:3
- Pk Info:3×1 mg
Specifications
About this item
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped at RT.
Caution: Research use only.