ContactUsLinkComponent
Portelite™ Rapid Fluorimetric Biotin Quantitation Kit
Portelite™ Rapid Fluorimetric Biotin Quantitation Kit
Catalog # AATB5539
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Portelite™ Rapid Fluorimetric Biotin Quantitation Kit
Catalog # AATB5539
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    50 Tests
Specifications
About this item
Optimised to use with CytoCite™ and Qubit™ fluorometers. Portelite™ rapid fluorimetric biotin quantitation kit is specifically optimised to quantify biotin and biotin conjugates with a portal fluorometer such as CytoCite™ and Qubit™ fluorometer.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
This fluorescence-based assay is much more sensitive than the commonly used colorimetric HABA assay.
Delivery: Shipped with blue ice.
Caution: Research use only.