ContactUsLinkComponent
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Catalog # AATB5537
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Amplite® Rapid Colorimetric Biotin Quantitation Kit
Catalog # AATB5537
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
Specifications
About this item
Optimised to use with Nanodrop. Amplite™ colorimetric biotin quantitation kit is designed to quantify the number of biotins attached to a protein or other macromolecule with a nanodrop spectrometer.
  • Absorption assessed at 500 nm
  • The kit is best used to determine biotin concentration in the range from 2 to 16 µM
  • 12-month shelf life
It is optimised to use with Nanodrop spectrometer.
Delivery: Shipped with blue ice.
Caution: Research use only.