Buccutite™ Streptavidin Antibody Conjugation Kit
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
Buccutite™ Streptavidin antibody conjugation kits are optimised for labelling either 25 µg, 100 µg or 1 mg of protein.
- The produced streptavidin-conjugated antibodies may be directly used in WB, ELISA, IHC without further purification
- Can also be used to conjugate other proteins or peptides
- It can be used to generate conjugates of different ratios of streptavidin/antibody
This streptavidin conjugation kit uses a simple and quick process for crosslinking streptavidin to an antibody.
Delivery: Shipped at RT.
Caution: Research use only.
Related Information
Manual Buccutite Streptavidin Antibody Conjugation Kit Optimized for Labeling 100 μg Protein.pdf Manual Buccutite Streptavidin Antibody Conjugation Kit Optimized for Labeling 25 μg Protein.pdf Manual Buccutite Streptavidin Antibody Conjµgation Kit Optimized for Labeling 100 µg Protein.pdf Manual Buccutite™ Streptavidin Antibody Conjugation Kit.PDF Manual Buccutite™ Streptavidin Antibody Conjµgation Kit Optimized For Labeling 1 Mg Protein.pdf
Manual
More
Frequently Bought Together