ContactUsLinkComponent
ReadiUse™ NADP Regenerating Kit
ReadiUse™ NADP Regenerating Kit
Artikelnr. AATB15266
Lieferant:  AAT BIOQUEST
New Product
CAS-Nummer:  
ReadiUse™ NADP Regenerating Kit
Artikelnr. AATB15266
Lieferant:  AAT BIOQUEST
New Product
CAS-Nummer:  
Spezifikationen
  • Pk:
    200 Tests
Spezifikationen
Über diesen Artikel
Designed for consistent and dependable results. AAT Bioquest's ReadiUse™ NADP regenerating kit provides a flexible prepackaged NADP regenerating system to accommodate various experimental designs.
  • Uses two ready-to-use solutions to regenerate NADP by a simple mixing
  • 300-500 enzyme assays can be performed using this kit
  • Six-month shelf life
This kit can be used for all NADP-requiring systems (such as: 12-Ketochenodeoxycholic Acid synthesis, Ferredoxin Reductase System).
Delivery: Shipped with blue ice.
Caution: Research use only.