ContactUsLinkComponent
Portelite™ Fluorimetric Protein Quantitation Kit
Portelite™ Fluorimetric Protein Quantitation Kit
Catalog # AATB11109
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Portelite™ Fluorimetric Protein Quantitation Kit
Catalog # AATB11109
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
Specifications
About this item
Optimised for CytoCite™ and Qubit™ fluorometers. The Portelite™ fluorimetric protein quantitation kit is significantly more sensitive than existing colorimetric protein measurements, e.g., Bradford and Bicinchoninic acid (BCA) assays.
  • Prolite™ Orange used in the kit is non-fluorescent in aqueous solution, but reacts rapidly with proteins and generates bright fluorescence
  • The assay has dynamic range from 12,5 µg/ml to 5 mg/ml of BSA
  • Six-month shelf life
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
Delivery: Shipped at RT.
Caution: Research use only.