Supelco® Organic Reference Standard, Pullulan Standard
Catalog # 18789-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Certified reference material, certified according to DIN, for GPC, 800000
- Description:Pullulan Standard
- Seq:24
- Storage conditions:2...8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:Not applicable