Transfectamine™ 6000 CRISPR Transfection Reagent
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
Transfectamine™ 6000 CRISPR Transfection reagent is a non-liposomal formula designed for optimal delivery and effectiveness of CRISPR/Cas9 components across a wide array of cell types.
- Consistently high transfection efficiency
- Higher viability of transfected cells
- Does not require special medium
Transfectamine™ 6000 CRISPR Transfection reagent is suitable for various applications, such as gene knockout and knock-in experiments, the generation of CRISPR-mediated gene-edited cell lines, functional genomics studies, and research in therapeutic gene editing.
Caution: For research use only.
Specifications
- Cell specific:Eukaryotic
- CRISPR/Cas 9:Yes
- Storage temperature:Freeze (< –15 °C); Minimise light exposure