Order Entry
Austria
ContactUsLinkComponent
Transfectamine™ 5000 Transfection Reagent
Transfectamine™ 5000 Transfection Reagent
Catalog # AATB60019
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Transfectamine™ 5000 Transfection Reagent
Catalog # AATB60019
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Cell specific:
    Eukaryotic
  • DNA and siRNA:
    Yes
  • DNA:
    Yes
  • dsRNA:
    Yes
  • shRNA:
    Yes
  • siRNA:
    Yes
  • Description:
    Transfectamine™ 5000 Transfection reagent is a powerful and versatile transfection reagent for the introduction of nucleic acids into eukaryotic cells, or more specifically, into animal cells
  • Storage temperature:
    Freeze (< –15 °C); Minimise light exposure

Specifications

About this item

Transfectamine™ 5000 Transfection Reagent is a powerful and versatile transfection reagent for the introduction of nucleic acids into eukaryotic cells, or more specifically, into animal cells.

  • Consistently high transfection efficiency
  • Higher viability of transfected cells
  • Does not require special medium

Transfectamine™ 5000 is easier to use compare to most other transfection reagents and does not require special medium.

Caution: For research use only.