ReadiLink™ Rapid XFD647 Antibody Labelling Kit
AATB5740
New Product
:
- Pk:3
- Pk Info:3×1 mg
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
: Shipped at RT.
: Research use only.