Order Entry
Austria
ContactUsLinkComponent
Conductivity Standard 2500 µS/cm at 25 °C
Conductivity Standard 2500 µS/cm at 25 °C
Catalog # ACSD5P53S.L5
Supplier:  CPACHEM
New Product
CAS Number:  
Conductivity Standard 2500 µS/cm at 25 °C
Catalog # ACSD5P53S.L5
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard
  • Conductivity:
    2500 µS/cm

Specifications

About this item

Certifications: ISO 17034, 17025, 9001 certified.