Supelco® TraceCERT® Organic Reference Standard, Metrafenone
Metrafenone CRM may also be used in the residue analysis of: 209 pesticides and persistent organic pollutants (POPs) extracted by QuEChERS-based procedure from fresh bovine and chicken liver samples using gas chromatography coupled to tandem mass spectrometry (GC-MS/MS). 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry. 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). 379 pesticides in human serum samples extracted by QuEChERS method using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Metrafenone in nine fresh vegetable samples using matrix solid-phase dispersion (MSPD) and HPLC-UV. Metrafenone in bitter gourd and soil samples using gas chromatography combined with an electron capture detector (GC-ECD).
: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
: P273 - P391 - P501
- Pk:50 mg
- Description:Metrafenone
- Cat. No.:18996-50MG
- M-Clarity quality level:MQ300
- Storage conditions:15...25 °C
- Hazard class description:H410
- MDL No.:MFCD08273825
- Synonym(s):(3-Bromo-6-methoxy-2-methylphenyl)(2,3,4-trimethoxy-6-methylphenyl)methanone; 3′-Bromo-2,3,4,6′-tetramethoxy-2′,6-dimethylbenzophenone
- Molecular weight:409,27
- CAS No.:220899-03-6
- Shipping conditions:STD
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



