Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, TIC Standard, 1000 ppm
Supelco® Organic Reference Standard, TIC Standard, 1000 ppm
Catalog # 12003-500ML-F
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, TIC Standard, 1000 ppm
Catalog # 12003-500ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    TIC Standard, 1000 ppm
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Concentration:
    1000 mg/L (nominal concentration (exact content on the CoA))

Specifications

About this item