Order Entry
Austria
ContactUsLinkComponent
Amplite® Tyramide (TSA) Signal Booster
Amplite® Tyramide (TSA) Signal Booster
Catalog # AATB44502
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
Amplite® Tyramide (TSA) Signal Booster
Catalog # AATB44502
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    1.000
  • Description:
    Amplite® Tyramide (TSA) signal booster
  • No. of reactions:
    1000

Specifications

About this item

Tyramide signal amplification (TSA) is a technique used to enhance the signal of target molecules, such as antibodies or nucleic acid probes, in immunohistochemistry and in situ hybridisation.

  • Stable for 12 months

Quality tested in-house.

Caution: For research use only.