Order Entry
Austria
ContactUsLinkComponent
Magicard™ CR80 PVC Cards with Magnetic Stripe and HoloPatch®
Magicard™ CR80 PVC Cards with Magnetic Stripe and HoloPatch®
Catalog # BRDY322003
Supplier:  Brady
New Product
undefined
Magicard™ CR80 PVC Cards with Magnetic Stripe and HoloPatch®
Catalog # BRDY322003
Supplier:  Brady
New Product

Specifications

  • Colour:
    White
  • Description:
    CR80 PVC cards with magnetic stripe and HoloPatch®
  • Length:
    86 mm
  • Material:
    PVC
  • Seq:
    05
  • Pk:
    500
  • Width:
    54 mm

Specifications

About this item

Plain white PVC card (CR80) with magnetic stripe and HoloPatch®.

  • Pre-approved for best quality printing on Magicard™ Printers
  • Magnetic stripe can be programmed with Magicard™ printers

Plain white PVC Card (CR80) with magnetic stripe and HoloPatch® for extra visual security. Cards are compatible with Magicard™ 300, Magicard™ 600, Pronto 100, and Prima 8 printers.

Packaging: 500 pieces per pack.