Order Entry
Austria
ContactUsLinkComponent
Cholesterol FS 10
Cholesterol FS 10
Catalog # DIAS113009910917
New Product
CAS Number:  
Cholesterol FS 10
Catalog # DIAS113009910917
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :386.66 g/mol
Density:1,0050 g/ml at 20 °C
Storage Temperature:Refrigerator
CAS Number:57-88-5
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646