Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Pullulan Standard
Supelco® Organic Reference Standard, Pullulan Standard
Catalog # 47053-25MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Pullulan Standard
Catalog # 47053-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Certified reference material
  • Description:
    Pullulan standard
  • Seq:
    18
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item