Supelco® Organic Reference Standard, ᴅ-(+)-Mannose
Catalog # 92683-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:3458-28-4
- Description:ᴅ-(+)-Mannose
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00064122
- M-Clarity quality level:MQ100
- Synonym(s):ᴅ-Mannopyranose
- Melting point:133 - 140 °C (lit.)
- Molecular weight:180,16