Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protonex™ Lyso-NIR 770
Protonex™ Lyso-NIR 770
Protonex™ Lyso-NIR 770
Artikelnr. AATB21242
Lieferant:
AAT BIOQUEST
New Product
CAS-Nummer:
Bild vergrößern
Protonex™ Lyso-NIR 770
Artikelnr. AATB21242
Lieferant:
AAT BIOQUEST
New Product
CAS-Nummer:
Spezifikationen
Pk:
1 mg
Seq:
2070
Spezifikationen
Über diesen Artikel
Produktdetails
:
:
736.76 g/mol
Produktdetails