Express Eingabe
Austria
AuftragsverfolgungKontakt
mFluor™ Violet 545 styramide
mFluor™ Violet 545 styramide
100 Slides, 100
Artikelnr. AATB45078
Anbieter:   AAT Bioquest
mFluor™ Violet 545 styramide
Artikelnr. AATB45078
Anbieter:  AAT Bioquest
CAS Nummer:  
mFluor™ Violet 545 styramide
100 Slides, 100
CAS Nummer:  
Preis
426,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
mFluor™ Violet 545 Styramide uses the bright green fluorescent dye mFluor™ Violet 545 (Ex/Em = 393/543 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Caution: For Research Use Only
Specifications
Specifications
  • VE:
    100
  • Pk Info:
    100 Slides
Specifications