Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2’,3’-cGAMP-iFluor 647
2’,3’-cGAMP-iFluor 647 conjugate
2’,3’-cGAMP-iFluor 647 conjugate
2’,3’-cGAMP-iFluor 647 conjugate
Catalog # AATB20322
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
2’,3’-cGAMP-iFluor 647 conjugate
Catalog # AATB20322
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
100 µG
Seq:
1286
Specifications
About this item
Product Details
:
:
1940.24 g/mol
Product Details