Anti-AGPAT5 Rabbit Polyclonal Antibody
ANTIA100620-100
New Product
- Antibody type:Primary
- Antigen name:1 acyl sn glycerol 3 phosphate acyltransferase epsilon
- Antigen symbol:AGPAT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9NUQ2
- Antigen synonyms:1-acylglycerol-3-phosphate O-acyltransferase 5|1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acid acyltransferase, epsilon)|1 AGPAT 5|1 AGP acyltransferase 5|1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon|1-AGPAT 5|1-AGP acyltransferase 5|1 AGPAT5|1 acylglycerol 3 phosphate O acyltransferase 5
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:42 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human AGPAT5 (amino acids 241-290)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to AGPAT5 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: AGPAT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human