Order Entry
Austria
ContactUsLinkComponent
 
Anti-SLC1A6 Rabbit Polyclonal Antibody
Catalog # ANTIA9109-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-SLC1A6 Rabbit Polyclonal Antibody
Catalog # ANTIA9109-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EAAT4
  • Antigen symbol:
    SLC1A6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    UniprotID# P48664
  • Antigen synonyms:
    High affinity neuronal glutamate transporter|MGC43671|Solute carrier family 1 (high affinity aspartate/glutamate transporter) member 6|Excitatory amino acid transporter 4|MGC33092|Solute carrier family 1 member 6|Sodium dependent glutamate/aspartate transporter
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,30, with 0,02% Sodium Azide and 50% Glycerol
  • Molecular weight:
    62kDa
  • Sequence:
    PGKGSKEGLHREGRIETIPTADAFMDLIRNMFPPNLVEACFKQFKTQYSTRVVTRTMVRTENGSEPGASMPPPFSVENGTSFLENVTRALGTLQEMLSFEETVPVPGSANG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 155-265 of human SLC1A6 (NP_001259016.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to SLC1A6 for WB with samples derived from Human, Mouse and Rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:100

Type: Primary
Antigen: SLC1A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat