Anti-CD169 Mouse Monoclonal Antibody [Clone: 7-239] (PE (Phycoerythrin))
Catalog # ANTIA86474-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD169
- Antigen symbol:CD169
- Clonality:Monoclonal
- Clone:7-239
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9BZZ2
- Antigen synonyms:FLJ00055|DKFZp667F058|FLJ32150|FLJ00411|dJ1009E24.1|FLJ00073|CD169 antigen|Sialic acid binding Ig like lectin 1|FLJ00051
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human rhinovirus 14-infected monocyte-derived dendritic cells
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [7-239] antibody to CD169 (PE) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD169
Clonality: Monoclonal
Clone: 7-239
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human