Order Entry
Austria
ContactUsLinkComponent
Anti-CD103 Mouse Monoclonal Antibody [Clone: Ber-ACT8] (FITC (Fluorescein Isothiocyanate))
Anti-CD103 Mouse Monoclonal Antibody [Clone: Ber-ACT8] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86443-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD103 Mouse Monoclonal Antibody [Clone: Ber-ACT8] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86443-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    A530055J10
  • Antigen symbol:
    CD103
  • Clonality:
    Monoclonal
  • Clone:
    Ber-ACT8
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P38570
  • Antigen synonyms:
    alpha-M290|aM290|alpha-E1|HML 1 antigen|CD 103|Antigen CD103|HML-1 antigen|CD103 antigen|HML1 antigen
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    HTLV-1 induced human T cell line MAPS16
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [Ber-ACT8] antibody to CD103 (FITC) for Flow Cytometry with samples derived from Human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: Ber-ACT8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates