ContactUsLinkComponent
EasySep™ Multistand with EasySep™ 'The Big Easy' Magnets
EasySep™ Multistand with EasySep™ 'The Big Easy' Magnets
Catalog # STMC18100
undefined
EasySep™ Multistand with EasySep™ 'The Big Easy' Magnets
Catalog # STMC18100
Specifications
  • Description:
    EasySep™ multistand with EasySep™ 'The Big Easy' magnets
  • Pk:
    1 KIT
Specifications
About this item
The EasySep™ Multistand allows the separation of up to four samples at one time.
  • Immunomagnetic column-free magnet
An ingenious reversible clamping bar allows the multistand to hold up to four EasySep™ magnets or four EasySep™ 'The Big Easy' magnets. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.
Packaging: This set includes the EasySep™ multistand and four EasySep™ 'The Big Easy' magnets.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').