Anti-FABP4 Mouse Monoclonal Antibody [clone: FABP4/4423]
ANTIA277587-100
New Product
- Antibody type:Primary
- Antigen name:3T3-L1 lipid-binding protein
- Antigen symbol:FABP4
- Clonality:Monoclonal
- Clone:FABP4/4423
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P15090
- Antigen synonyms:adipocyte|422/aP2|ALBP|Adipocyte lipid binding protein|A-FABP|Adipocyte lipid-binding protein|Adipocyte protein AP2|AFABP|Adipocyte-type fatty acid-binding protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:15 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1-132, of human FABP4 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [FABP4/4423] antibody to FABP4 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: FABP4
Clonality: Monoclonal
Clone: FABP4/4423
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human