Order Entry
Austria
ContactUsLinkComponent
Anti-FABP4 Mouse Monoclonal Antibody [Clone: FABP4/4424]
Anti-FABP4 Mouse Monoclonal Antibody [Clone: FABP4/4424]
Catalog # ANTIA277588-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-FABP4 Mouse Monoclonal Antibody [Clone: FABP4/4424]
Catalog # ANTIA277588-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3T3-L1 lipid-binding protein
  • Antigen symbol:
    FABP4
  • Clonality:
    Monoclonal
  • Clone:
    FABP4/4424
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P15090
  • Antigen synonyms:
    adipocyte|422/aP2|ALBP|Adipocyte lipid binding protein|A-FABP|Adipocyte lipid-binding protein|Adipocyte protein AP2|AFABP|Adipocyte-type fatty acid-binding protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    15 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 1-132, of human FABP4 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [FABP4/4424] antibody to FABP4 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: FABP4
Clonality: Monoclonal
Clone: FABP4/4424
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human