Anti-APOA1BP Rabbit Polyclonal Antibody
ANTIA96421-100
New Product
- Antibody type:Primary
- Antigen name:AI BP
- Antigen symbol:APOA1BP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q8NCW5
- Antigen synonyms:APOA1BP|NAD(P)HX epimerase|Apolipoprotein A I binding protein|ApoA I binding protein|MGC119143|NAD(P)H hydrate epimerase|MGC119144|AIBP|MGC119145
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:31 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human AIBP (amino acids 121-170)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to AIBP for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: APOA1BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat