Order Entry
Austria
ContactUsLinkComponent
Anti-UAP56 Rabbit Monoclonal Antibody [clone: ARC1731]
Anti-UAP56 Rabbit Monoclonal Antibody [clone: ARC1731]
Catalog # ANTIA308352-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-UAP56 Rabbit Monoclonal Antibody [clone: ARC1731]
Catalog # ANTIA308352-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    0610030D10Rik
  • Antigen symbol:
    UAP56
  • Clonality:
    Monoclonal
  • Clone:
    ARC1731
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q13838
  • Antigen synonyms:
    B(0,+)-type amino acid transporter 1|D17H6S81E-1|D17H6S81E|ATP-dependent RNA helicase p47|56 kDa U2AF65-associated protein|BAT1|AI428441|4F2-LC6|Bat1a
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    49 kDa
  • Sequence:
    QNFPAIAIHRGMPQEERLSRYQQFKDFQRRILVATNLFGRGMDIERVNIAFNYDMPEDSDTYLHRVARAGRFGTKGLAITFVSDENDAKILNDVQDRFEVNISELPDEIDISSYIEQTR
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 310-428 of human UAP56/DDX39B (Q13838)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC1731] antibody to UAP56 for WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200

Type: Primary
Antigen: UAP56
Clonality: Monoclonal
Clone: ARC1731
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat