Orders LinkContactUsLinkComponent
CryoPlus™ Sample Storage Systems
CryoPlus™ Sample Storage Systems
CryoPlus™ Sample Storage Systems
Certificates
CryoPlus™ Sample Storage Systems
7 Options Available Below
Limited Time Sale
Select items below are currently on sale
Product Details & Documents
Increase valuable laboratory storage space with CryoPlus™ Storage Systems, which store up to 38500 vials (2,0 ml). The ideal combination of liquid nitrogen, storage reliability and microprocessor technology, the CryoPlus series LN₂ storage system provides precise and accurate control over all parameters without complicated programming.
  • 16 preset audible and visual alarm combinations provide maximum product protection
  • Automatic defogger incorporated into the manual fill index to improve visibility during retrieval
  • Liquid level sensor and fill port within vacuum space, for maximum storage and sensor protection
  • Front-mounted key lock and rear-mounted recessed power switch; control panel can also be locked
  • Temperature sleeve (included) provides colder temperatures and efficient vapour phase operation
The units feature advanced microprocessor control for precise liquid level management, accommodating over 16 parameters, and include an autofill system for seamless operation. The control panel, conveniently mounted on top, offers easy access and touchpad programming. Under the lid, a digital temperature display sensor indicates the highest temperature reading inside the tank. The tank's status is continuously monitored by 24 tricolour LEDs that display the actual liquid nitrogen level and high/low-level setpoints. Alarm contacts are available for remote monitoring of storage tank conditions. Constructed with durability in mind, the low-profile tank boasts a vacuum-insulated, stainless-steel chamber and is on heavy-duty casters for mobility.
The counterbalanced lid, featuring foamed-in-place, high-density polyurethane insulation and a dedicated vent, ensures 100% clearance. Additionally, two independent, flexible lid gaskets minimise moisture migration into the chamber. The system is designed to accommodate 2 ml vials based on a specific rack configuration, ensuring efficient storage and retrieval.
Certifications: CFDA, cULUS
Ordering information: Optional racks, canisters, frames and risers allow you to create an organised inventory system that provides optimum storage in liquid or vapour phase.
Specifications for Product Family
Specifications
  • Catalog No:
  • THBR7401GP
  • THBR7403GP
  • BARN7405
  • THBR7405GP
  • BARN7407
  • THBR7407GP
  • BARN7401
  • Model:
  • CryoPlus 1
  • CryoPlus 2
  • CryoPlus 3
  • CryoPlus 3
  • CryoPlus 4
  • CryoPlus 4
  • CryoPlus 4
  • Evaporation rate:
  • ~ 4 L/d
  • ~ 6 L/d
  • ~ 9 L/d
  • ~ 9 L/d
  • ~ 12 L/d
  • ~ 12 L/d
  • ~ 12 L/d
  • Int.Ø:
  • 406 mm
  • 610 mm
  • 787 mm
  • 787 mm
  • 1103 mm
  • 1103 mm
  • 1103 mm
  • Vial capacity:
  • 5500 - 6318
  • 13000
  • 22100 - 24000
  • 22100 - 24000
  • 36400 - 38500
  • 36400 - 38500
  • 36400 - 38500
  • Weight Empty:
  • 97,1 kg
  • 147,4 kg
  • 188,7 kg
  • 188,7 kg
  • 371,9 kg
  • 371,9 kg
  • 371,9 kg
  • Weight Full:
  • 160,6 kg
  • 308,4 kg
  • 463,1 kg
  • 463,1 kg
  • 734,8 kg
  • 734,8 kg
  • 734,8 kg
Specifications
Product Family Options
Product Information
  • Description
    Verfügbarkeit
    Preis
  • CryoPlus 1 sample storage system, 90 L, 660L×546W×1040H mm, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10 754,00 €
    10 754,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 1 ST
       10 754,00 €
    Specifications:
    New Product
    Mehr
    Specifications
    Cat. No.
    THBR7401GP
    Model
    CryoPlus 1
    Evaporation rate
    ~ 4 L/d
    Int.Ø
    406 mm
    Vial capacity
    5500 - 6318
    Weight Empty
    97,1 kg
    Weight Full
    160,6 kg
    New Product
  • CryoPlus 2 sample storage system, 200 L, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13 910,00 €
    13 910,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 1 ST
       13 910,00 €
    Specifications:
    New Product
    Mehr
    Specifications
    Cat. No.
    THBR7403GP
    Model
    CryoPlus 2
    Evaporation rate
    ~ 6 L/d
    Int.Ø
    610 mm
    Vial capacity
    13000
    Weight Empty
    147,4 kg
    Weight Full
    308,4 kg
    New Product
  • CryoPlus 3 sample storage system, 340 L, 1054L×876W×1041H mm, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30 450,00 €
    30 450,00 €
    1 * 1 ST
       30 450,00 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BARN7405
    Model
    CryoPlus 3
    Evaporation rate
    ~ 9 L/d
    Int.Ø
    787 mm
    Vial capacity
    22100 - 24000
    Weight Empty
    188,7 kg
    Weight Full
    463,1 kg
  • CryoPlus 3 sample storage system, 340 L, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19 500,00 €
    19 500,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 1 ST
       19 500,00 €
    Specifications:
    New Product
    Mehr
    Specifications
    Cat. No.
    THBR7405GP
    Model
    CryoPlus 3
    Evaporation rate
    ~ 9 L/d
    Int.Ø
    787 mm
    Vial capacity
    22100 - 24000
    Weight Empty
    188,7 kg
    Weight Full
    463,1 kg
    New Product
  • CryoPlus 4 sample storage system, 552 L, 1270L×1105W×1194H mm, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43 140,00 €
    43 140,00 €
    1 * 1 ST
       43 140,00 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BARN7407
    Model
    CryoPlus 4
    Evaporation rate
    ~ 12 L/d
    Int.Ø
    1103 mm
    Vial capacity
    36400 - 38500
    Weight Empty
    371,9 kg
    Weight Full
    734,8 kg
  • CryoPlus 4 sample storage system, 552 L, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27 625,00 €
    27 625,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 1 ST
       27 625,00 €
    Specifications:
    New Product
    Mehr
    Specifications
    Cat. No.
    THBR7407GP
    Model
    CryoPlus 4
    Evaporation rate
    ~ 12 L/d
    Int.Ø
    1103 mm
    Vial capacity
    36400 - 38500
    Weight Empty
    371,9 kg
    Weight Full
    734,8 kg
    New Product
  • CryoPlus 4 sample storage system, 552 L, 208 - 230 V, 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    Mehr
    Specifications
    Cat. No.
    BARN7401
    Model
    CryoPlus 4
    Evaporation rate
    ~ 12 L/d
    Int.Ø
    1103 mm
    Vial capacity
    36400 - 38500
    Weight Empty
    371,9 kg
    Weight Full
    734,8 kg
    New Product
Accessories
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • BARN1950834
    Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000001
    Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials 607×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 478,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • BARN1950840
    Rack for Matrix 2D plates measuring 58,42 mm tall, 699×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
    Log in for availability
    1 * 1 ST - 170,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR195701
    Field installed (QI) 220 V, 50/60 Hz, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 3 500,00 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000371
    Frame for HERA4000335 canister, 602×15×165 mm
    CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 129,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000060
    88,9 mm riser, for CryoPlus 1, 2
    Log in for availability
    1 * 1 ST - 391,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000066
    88,9 mm riser, for CryoPlus 4
    Log in for availability
    1 * 1 ST - 694,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR195023
    Factory installed 220 V, 50/60 Hz, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 2 060,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000002
    Rack includes stainless steel boxes for 1,5/2 ml vials, 607×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 932,00 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000112
    Arrowhead rack, liquid phase, 1,5/2 ml capacity, includes 12 cardboard boxes with 81 cell divider, holds 12 boxes/rack, 660×198×191 mm
    CryoPlus 1
    Log in for availability
    1 * 1 ST - 961,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000061
    127 mm riser, for CryoPlus 1, 2
    Log in for availability
    1 * 1 ST - 414,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • HERA4000369-MAR
    Frame for Gambro DF-700 canisters, 671×25×254 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 89,20 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000113
    Arrowhead rack, liquid phase, 1,5/2 ml capacity, includes 12 stainless steel boxes with 81 cell divider, holds 12 boxes/rack, 660×198×191 mm
    CryoPlus 1
    Log in for availability
    1 * 1 ST - 750,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • HERA4000290
    T-valve assembly with fittings, used to connect two transfer hoses to a supply tank for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 401,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • HERA189470
    Mini cardboard box, 69,85×69,85×50,8 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 13,80 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000013
    Cardboard divider, 5 ml capacity, 100-cell
    Square cryo box, CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 8,30 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000006
    Square rack, liquid phase, 1,5/2 ml capacity, holds 13 cardboard boxes with 100 cell divider, holds 13 boxes/rack, 696×140×143 mm
    CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 585,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000007
    Rack includes stainless steel boxes and cardboard dividers for 1,5/2 ml vials, 696×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 855,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000042
    Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 660×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 650,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000008
    Rack includes cardboard boxes and cardboard dividers for 4 ml vials, 645×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 433,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000010
    Rack includes cardboard boxes and cardboard dividers for 5 ml vials, 676×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 364,00 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000336
    Sliding canister for 500 ml bag, Fenwal 4R5462 or OriGen CS 500, 142×234×10 mm
    CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 79,20 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • MATR1950839
    Rack for Matrix 2D plates measuring 39,37 - 54,61 mm tall, 690×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000044
    Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 714×198×191 mm, for CryoPlus 1
    Log in for availability
    1 * 1 ST - 961,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000067
    127 mm riser, for CryoPlus 4
    Log in for availability
    1 * 1 ST - 754,00 €
Product Information
  • Artikelnr.
    Description
    For
    Verfügbarkeit
    Preis
  • HERA4000238
    Stainless steel boxes, 1,5/2 ml (without 100 cell divider)
    CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 36,50 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR4000003
    Rack includes cardboard boxes and cardboard dividers for 4 ml vials, 566×140×143 mm, for CryoPlus 1, 2, 4
    Log in for availability
    1 * 1 ST - 387,00 €
Product Information
  • Artikelnr.
    Description
    Verfügbarkeit
    Preis
  • THBR1950707
    Extend inner sleeve, CP2
    Log in for availability
    1 * 1 ST - 350,00 €