Anti-PML+RARA Fusion Rabbit Polyclonal Antibody
Catalog # ANTIA99234-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:MYL
- Antigen symbol:PML+RARA Fusion
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P29590
- Antigen synonyms:Promyelocytic leukemia protein|Promyelocytic leukemia|PML+RARA Fusion|Promyelocytic leukemia, inducer of|PP8675|NR1B1|Nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form|RAR|Nuclear receptor subfamily 1 group B member 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:97 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PML (amino acids 11-60)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to PML for WB, IHC, IF and ELISA with samples derived from human.
- Validated applications: WB, IHC, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: PML+RARA Fusion
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human