Anti-EDG6 Rabbit Polyclonal Antibody
Artikelnr. ANTIA98408-100
Lieferant: ANTIBODIES.COM
New Product
Spezifikationen
- Antibody type:Primary
- Antigen name:EDG6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O95977
- Antigen synonyms:Putative G protein coupled receptor|MGC129298|Endothelial differentiation G protein coupled receptor 6|S1P receptor 4|Lysophospholipid receptor C1|Endothelial differentiation gene 6|Endothelial differentiation G-protein coupled receptor 6|LPC1|Endothelial differentiation lysophosphatidic acid G protein coupled receptor 6
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human EDG6 (amino acids 326-375)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Spezifikationen
Über diesen Artikel
Rabbit polyclonal antibody to EDG6 for WB, IF and ELISA with samples derived from human and mouse.
- Validated applications: WB, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: EDG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse