Order Entry
Austria
ContactUsLinkComponent
Anti-BCKDK Rabbit Monoclonal Antibody [clone: ARC2875]
Anti-BCKDK Rabbit Monoclonal Antibody [clone: ARC2875]
Catalog # ANTIA307436-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-BCKDK Rabbit Monoclonal Antibody [clone: ARC2875]
Catalog # ANTIA307436-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCKD kinase
  • Antigen symbol:
    BCKDK
  • Clonality:
    Monoclonal
  • Clone:
    ARC2875
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O14874
  • Antigen synonyms:
    Branched chain ketoacid dehydrogenase kinase|Bckdk|BCKDKD|BCKD_HUMAN|BCKDH kinase|BCKD-kinase|BCKDHKIN|Branched chain alpha keto acid dehydrogenase kinase|BDK
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    46 kDa
  • Sequence:
    MILASVLRSGPGGGLPLRPLLGPALALRARSTSATDTHHVEMARERSKTVTSFYNQSAIDAAAEKPSVRLTPTMMLYAGRSQDGSHLLKSARYLQQELPV
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BCKDK (O14874).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC2875] antibody to BCKDK for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: BCKDK
Clonality: Monoclonal
Clone: ARC2875
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat