Orders LinkContactUsLinkComponent
CytoCite™ Sample Tubes
CytoCite™ Sample Tubes
500 μl tubes, CytoCite™ sample tubes
Catalog # AATBCCT100
Supplier:   AAT Bioquest
CytoCite™ Sample Tubes
Catalog # AATBCCT100
Supplier:  AAT Bioquest
CytoCite™ Sample Tubes
500 μl tubes, CytoCite™ sample tubes
Price
138,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Cytocite™ sample tubes are 500 μl thin-walled polypropylene tubes validated for use with Cytocite or Qubit Fluorometer.
  • Storage at room temperature
Optimised for easy use with both common fluorometer instruments.
Caution: Research use only.
Specifications
  • Capacity:
    500 μl tubes
  • Description:
    CytoCite™ sample tubes