ContactUsLinkComponent
ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Catalog # AATB7510
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Catalog # AATB7510
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    ReadiCleave™ AML cleavage buffer 5X aqueous solution
Specifications
About this item
ReadiCleave™ AML Cleavage Buffer provides an optimised formula to effectively remove the fluorescence tags from ReadiCleave™ AML-labeled biological targets when needed.
  • Designed for ease of use
  • Saves time
The removal of the fluorescent ReadiCleave™ AML tags may make the fluorescence imaging more compatible with the downstream analysis of a biological target by other techniques, e.g., MS analysis.
Caution: For research use only.