Order Entry
Austria
ContactUsLinkComponent
10X Citrate Buffer, pH 6,0
10X Citrate Buffer, pH 6,0
Catalog # AATB10000
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
10X Citrate Buffer, pH 6,0
Catalog # AATB10000
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    10X citrate buffer pH 6.0

Specifications

About this item

A citrate buffer is an aqueous solution composed of a weak acid (citric acid) and its conjugate base (sodium citrate).

  • Designed for ease of use
  • Saves time

Buffer solution is intended for heat-induced antigen retrieval in IHC.

Caution: For research use only.