Anti-IgG Goat Polyclonal Antibody (iFluor® 860)
New Product
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Purified and quality-tested for consistent results
- Suitable for multiple applications
- Purified through affinity chromatography
iFluor® 860 goat anti-rabbit IgG (H+L) conjugates are ideal for various NIR imaging applications, including Western blotting, ELISA, protein arrays, tissue section imaging, and in vivo imaging.
: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 860
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
- Catalog No:
- AATB48069
- AATB48068
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- iFluor® 860
- iFluor® 860
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Goat
- Goat
- Isotype:
- IgG (H, L)
- IgG (H, L)
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- Minimal cross reaction to human, horse, rabbit and bovine
- Minimal cross reaction to human, horse, rabbit and bovine
- Storage temperature:
- Freeze (<−15 °C)
- Freeze (<−15 °C)
- Shipping temperature:
- 20
- 20
Frequently Bought Together