Anti-SIGLEC1 Mouse Monoclonal Antibody [clone: HSn 7D2]
Catalog # ANTIA253169-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD169
- Antigen symbol:SIGLEC1
- Clonality:Monoclonal
- Clone:HSn 7D2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9ZZ2
- Antigen synonyms:FLJ00055|DKFZp667F058|FLJ32150|FLJ00411|dJ1009E24.1|FLJ00073|CD169 antigen|Sialic acid binding Ig like lectin 1|FLJ00051
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:185 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fc fusion protein containing N-terminal 4 domains of human sialoadhesin
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HSn 7D2] antibody to SIGLEC1 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: SIGLEC1
Clonality: Monoclonal
Clone: HSn 7D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human