Order Entry
Austria
ContactUsLinkComponent
Anti-CD170 Mouse Monoclonal Antibody [clone: 1A5] (PE (Phycoerythrin))
Anti-CD170 Mouse Monoclonal Antibody [clone: 1A5] (PE (Phycoerythrin))
Catalog # ANTIA121863-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD170 Mouse Monoclonal Antibody [clone: 1A5] (PE (Phycoerythrin))
Catalog # ANTIA121863-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD170
  • Antigen symbol:
    CD170
  • Clonality:
    Monoclonal
  • Clone:
    1A5
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O15389
  • Antigen synonyms:
    OBBP2|Sialic acid binding Ig-like lectin 5|OB-binding protein 2|mSiglec-F|OB-BP2|CD33L2|Obesity-binding protein 2|CD33 antigen-like 2|CD170 antigen
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Extracellular domain of human CD170 fused to human IgG1 (Fc)
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [1A5] antibody to CD170 (PE) for Flow Cytometry with samples derived from human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD170
Clonality: Monoclonal
Clone: 1A5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates