Order Entry
Austria
ContactUsLinkComponent
Anti-ADCY8 / AC8 Mouse Monoclonal Antibody [clone: ADCY8/7341]
Anti-ADCY8 / AC8 Mouse Monoclonal Antibody [clone: ADCY8/7341]
Catalog # ANTIA315820-100
Supplier:  ANTIBODIES.COM
New Product
Anti-ADCY8 / AC8 Mouse Monoclonal Antibody [clone: ADCY8/7341]
Catalog # ANTIA315820-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADCY8 / AC8
  • Clonality:
    Monoclonal
  • Clone:
    ADCY8/7341
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P40145
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    140 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 1-200) of human ADCY8 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [ADCY8/7341] antibody to ADCY8 / AC8 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: ADCY8 / AC8
Clonality: Monoclonal
Clone: ADCY8/7341
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human